Anti-FGR Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: FGR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77855-411
- 77737-188
- Antigen Symbol:
- FGR
- FGR
- Antigen Synonyms:
- c c fgr protooncogene|c src 2 proto oncogene|c src2|FGR_HUMAN|FLJ43153|Gardner Rasheed feline sarcoma viral (v fgr)|Gardner Rasheed feline sarcoma viral (v fgr) oncogene homolog|Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog|MGC75096|p55 c fgr protein|P55 P55-p55c p58-p58c p58c-Proto oncogene c Proto oncogene tyrosine protein kinase Proto-oncogene c-SRC 2|SRC2|Tyrosine protein kinase Tyrosine-protein kinase Fgr
- c c fgr protooncogene|c src 2 proto oncogene|c src2|FGR_HUMAN|FLJ43153|Gardner Rasheed feline sarcoma viral (v fgr)|Gardner Rasheed feline sarcoma viral (v fgr) oncogene homolog|Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog|MGC75096|p55 c fgr protein|P55 P55-p55c p58-p58c p58c-Proto oncogene c Proto oncogene tyrosine protein kinase Proto-oncogene c-SRC 2|SRC2|Tyrosine protein kinase Tyrosine-protein kinase Fgr
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09769
- P09769
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Tyrosine-protein kinase Fgr protein (11-72AA)
- Recombinant human Tyrosine-protein kinase Fgr protein (11-72AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice