Anti-USP24 Rabbit Polyclonal Antibody
77736-790
New Product
- Antigen Symbol:USP24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UPU5
- Antigen Synonyms:Ubiquitin thioesterase 24|Ubiquitin thiolesterase 24|Ubiquitin-specific-processing protease 24|Ubiquitin specific protease 24|Ubiquitin carboxyl-terminal hydrolase 24|Ubiquitin carboxyl terminal hydrolase 24|Ubiquitin specific processing protease 24|UBP24_HUMAN|KIAA1057|Ubiquitin specific peptidase 24|Deubiquitinating enzyme 24
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Deubiquitinating enzyme 24
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: USP24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse