Anti-MPPED1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: MPPED1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77743-340
- 77859-910
- Antigen Symbol:
- MPPED1
- Antigen Synonyms:
- MPPED1|C22orf1|FAM1A|Metallophosphoesterase domain-containing protein 1|EC 3.1.-.-|Adult brain protein 239|239AB
- MPPED1|C22orf1|FAM1A|Metallophosphoesterase domain-containing protein 1|EC 3.1.-.-|Adult brain protein 239|239AB
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O15442
- O15442
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Metallophosphoesterase domain-containing protein 1 protein (1-326AA)
- Recombinant Human Metallophosphoesterase domain-containing protein 1 protein (1-326AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice