Anti-KRT81 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT81
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-907
- 77858-772
- Antigen Symbol:
- KRT81
- Antigen Synonyms:
- 1|basic|ghHb 1|ghHb1|ghHkb 1|ghHKb1|hair|Hair keratin K2.9|Hard keratin type II 1|HB 1|HB1|hHAKB2 1|K2.9|K81|Keratin 81|Keratin|Keratin hair basic 1|Keratin type II cuticular Hb1|Keratin-81|Keratin81|KRT 81|KRT81|KRT81_HUMAN|KRTHB 1|KRTHB1|Metastatic lymph node 137 gene protein|MLN 137|MLN137|type II cuticular Hb1|Type II hair keratin Hb1|Type-II keratin Kb21
- 1|basic|ghHb 1|ghHb1|ghHkb 1|ghHKb1|hair|Hair keratin K2.9|Hard keratin type II 1|HB 1|HB1|hHAKB2 1|K2.9|K81|Keratin 81|Keratin|Keratin hair basic 1|Keratin type II cuticular Hb1|Keratin-81|Keratin81|KRT 81|KRT81|KRT81_HUMAN|KRTHB 1|KRTHB1|Metastatic lymph node 137 gene protein|MLN 137|MLN137|type II cuticular Hb1|Type II hair keratin Hb1|Type-II keratin Kb21
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q14533
- Q14533
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Keratin, type II cuticular Hb1 protein (3-505AA)
- Recombinant Human Keratin, type II cuticular Hb1 protein (3-505AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice