Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MOCS2 Rabbit Recombinant Polyclonal Antibody
Anti-MOCS2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MOCS2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77859-875
  • 77743-291
  • Antigen Symbol:
  • MOCS2
  • MOCS2
  • Antigen Synonyms:
  • MCBPE|MOCO1|Molybdopterin synthase catalytic subunit|EC 2.8.1.12|MOCO1-B|Molybdenum cofactor synthesis protein 2 large subunit|Molybdenum cofactor synthesis protein 2B|MOCS2B|Molybdopterin-synthase large subunit|MPT synthase large subunit
  • EC 2.8.1.12|MOCO1|MOCO1-B|Molybdenum cofactor synthesis protein 2 large subunit|Molybdopterin synthase catalytic subunit|Molybdenum cofactor synthesis protein 2B|MOCS2B|MCBPE|Molybdopterin-synthase large subunit|MPT synthase large subunit
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O96007
  • O96007
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Molybdopterin synthase catalytic subunit protein (1-188AA)
  • Recombinant Human Molybdopterin synthase catalytic subunit protein (1-188AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications