Anti-HIP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:200 to 1:1000, IHC:1:20 to 1:200
Type:
Antigen: HIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-692
- 77856-855
- Antigen Symbol:
- HIP1
- HIP1
- Antigen Synonyms:
- 2610109B09Rik|A930014B11Rik|E130315I21Rik|HIP 1|HIP I|HIP-1|HIP-I|hip1|HIP1/PDGFRB fusion gene|HIP1/PDGFRB fusion gene, included|HIP1_HUMAN|HIPI|Huntingtin interacting protein 1|Huntingtin Interacting Protein HIP1|Huntingtin-interacting protein 1|Huntingtin-interacting protein I|ILWEQ|KIAA4113|MGC126506|MGC27616|mKIAA4113
- 2610109B09Rik| A930014B11Rik| E130315I21Rik| HIP 1| HIP I| HIP-1| HIP-I| hip1| HIP1/PDGFRB fusion gene| HIP1/PDGFRB fusion gene, included| HIP1_HUMAN| HIPI| Huntingtin interacting protein 1| Huntingtin Interacting Protein HIP1| Huntingtin-interacting protein 1| Huntingtin-interacting protein I| ILWEQ| KIAA4113| MGC126506| MGC27616| mKIAA4113
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00291
- O00291
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Huntingtin-interacting protein 1 protein (410-690AA)
- Recombinant Human Huntingtin-interacting protein 1 protein (410-690AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice