Anti-GRP Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-490
- 77739-077
- Antigen Symbol:
- GRP
- Antigen Synonyms:
- BN|Bombesin|GRP|GRP-10|GRP_HUMAN|GRP10|Neuromedin-C|NeuromedinC|Pre progastrin releasing peptide|PreproGRP|ProGRP
- BN|Bombesin|GRP|GRP-10|GRP_HUMAN|GRP10|Neuromedin-C|NeuromedinC|Pre progastrin releasing peptide|PreproGRP|ProGRP
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P07492
- P07492
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Gastrin-releasing peptide protein (31-98AA)
- Recombinant Human Gastrin-releasing peptide protein (31-98AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice