Anti-ZDHHC15 Rabbit Polyclonal Antibody
77735-810
New Product
- Antigen Symbol:ZDHHC15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96MV8
- Antigen Synonyms:UNQ1969/PRO4501|Palmitoyltransferase EC 2.3.1.225|Zinc finger DHHC domain-containing protein 15|DHHC-15
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:DHHC-15
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:10000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human