Anti-GPS1 Rabbit Polyclonal Antibody
Catalog # 77738-922
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GPS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13098
- Antigen Synonyms:COP9 signalosome complex subunit 1|COP9 signalosome subunit 1|COPS1|CSN 1|CSN1_HUMAN|FUS6/COP11|Arabidopsis|homolog of|G protein pathway suppressor 1|GPS-1|JAB1-containing signalosome subunit 1|Protein Protein MFH|SGN1|Signalosome subunit 1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Signalosome subunit 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat