Anti-GIMAP4 Rabbit Polyclonal Antibody
77738-258
New Product
- Antigen Symbol:GIMAP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q9NUV9
- Antigen Synonyms:FLJ11110|GIMA4_HUMAN|Gimap4|GTPase IMAP family member 4|hIAN1|HIMAP4|Human immune associated nucleotide 1|IAN-1|IAN1|IMAP4|Immunity associated nucleotide 1 protein|Immunity associated protein 4|Immunity-associated nucleotide 1 protein|Immunity-associated protein 4|MSTP062
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MSTP062
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: GIMAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human