Anti-ITM2B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ITM2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-359
- 77858-349
- Antigen Symbol:
- ITM2B
- ITM2B
- Antigen Synonyms:
- ABRI|ABri/ADan amyloid peptide|BRI 2|BRI|BRI2|BRICD 2B|BRICD2B|BRICHOS domain containing 2B|E25B|E3 16|E3-16|FBD|imBRI2|Immature BRI2|Integral membrane protein 2B|ITM 2B|ITM2B|ITM2B_HUMAN|Protein E25B|RDGCA|Transmembrane protein BRI
- ABRI|ABri/ADan amyloid peptide|BRI 2|BRI|BRI2|BRICD 2B|BRICD2B|BRICHOS domain containing 2B|E25B|E3 16|E3-16|FBD|imBRI2|Immature BRI2|Integral membrane protein 2B|ITM 2B|ITM2B|ITM2B_HUMAN|Protein E25B|RDGCA|Transmembrane protein BRI
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y287
- Q9Y287
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Integral membrane protein 2B protein (81-148AA)
- Recombinant Human Integral membrane protein 2B protein (81-148AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice