Anti-MAP1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-353
- 77742-622
- Antigen Symbol:
- MAP1A
- MAP1A
- Antigen Synonyms:
- MAP 1A|MAP 1L|MAP-1A|MAP1 light chain LC2|MAP1A_HUMAN|MAP1L|Microtubule associated protein 1 like|Microtubule associated protein 1A|MTAP 1A|MTAP1A|Proliferation related protein p80|Proliferation-related protein p80
- MAP 1A|MAP 1L|MAP-1A|MAP1 light chain LC2|MAP1A_HUMAN|MAP1L|Microtubule associated protein 1 like|Microtubule associated protein 1A|MTAP 1A|MTAP1A|Proliferation related protein p80|Proliferation-related protein p80
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78559
- P78559
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Microtubule-associated protein 1A protein (1979-2168AA)
- Recombinant Human Microtubule-associated protein 1A protein (1979-2168AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice