Anti-MLPH Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MLPH
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-827
- 77743-222
- Antigen Symbol:
- MLPH
- MLPH
- Antigen Synonyms:
- 2210418F23Rik|5031433I09Rik|AW228792|D1Wsu84e|Exophilin 3|Exophilin-3|l(1)-3Rk|l1Rk3|Leaden|Leaden protein|ln|Melanophilin|MELPH_HUMAN|MGC2771|MGC59733|Slac 2a|SlaC2-a|Slp homolog lacking C2 domains a|Synaptotagmin like protein 2a|Synaptotagmin like protein lacking C2 domains A|Synaptotagmin-like protein 2a
- 2210418F23Rik| 5031433I09Rik| AW228792| D1Wsu84e| Exophilin 3| Exophilin-3| l(1)-3Rk| l1Rk3| Leaden| Leaden protein| ln| Melanophilin| MELPH_HUMAN| MGC2771| MGC59733| Slac 2a| SlaC2-a| Slp homolog lacking C2 domains a| Synaptotagmin like protein 2a| Synaptotagmin like protein lacking C2 domains A| Synaptotagmin-like protein 2a
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BV36
- Q9BV36
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Melanophilin protein (310-539AA)
- Recombinant Human Melanophilin protein (310-539AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice