Anti-Igfbp1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: Igfbp1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Specifications
- Catalog No:
- 77740-752
- 77857-931
- Antigen Symbol:
- Igfbp1
- Igfbp1
- Antigen Synonyms:
- Igfbp-1|Insulin-like growth factor-binding protein 1|IBP-1|IGF-binding protein 1
- Igfbp-1|Insulin-like growth factor-binding protein 1|IBP-1|IGF-binding protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse, Human
- Mouse, Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P47876
- P47876
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant mouse Insulin-like growth factor-binding protein 1 protein (26-272AA)
- Recombinant mouse Insulin-like growth factor-binding protein 1 protein (26-272AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice