Anti-GRB10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRB10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-420
- 77738-980
- Antigen Symbol:
- GRB10
- GRB10
- Antigen Synonyms:
- grb 10| GRB IR| grb-10| GRB10 adapter protein| GRB10 adaptor protein| GRB10| GRB10_HUMAN| GRBIR| Growth factor receptor bound protein 10| Growth factor receptor-bound protein 10| Insulin receptor binding protein| Insulin receptor binding protein GRB IR| Insulin receptor-binding protein Grb-IR| IRBP| KIAA0207| Maternally expressed gene 1| MEG1| RSS
- grb 10|GRB IR|grb-10|GRB10 adapter protein|GRB10 adaptor protein|GRB10|GRB10_HUMAN|GRBIR|Growth factor receptor bound protein 10|Growth factor receptor-bound protein 10|Insulin receptor binding protein|Insulin receptor binding protein GRB IR|Insulin receptor-binding protein Grb-IR|IRBP|KIAA0207|Maternally expressed gene 1|MEG1|RSS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13322
- Q13322
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Growth factor receptor-bound protein 10 protein (15-180AA)
- Recombinant Human Growth factor receptor-bound protein 10 protein (15-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice