Anti-MOB1A Rabbit Polyclonal Antibody
Catalog # 77743-279
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MOB1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9H8S9
- Antigen Synonyms:B1|Mps One Binder kinase activator like 1B (yeast)|C2orf6|Chromosome 2 open reading frame 6|FLJ10788|FLJ11595|MATS1|MOB kinase activator 1A|MOB1|Mob1 alpha|Mob1 homolog 1B|MOB1 Mps One Binder homolog A|MOB1 Mps One Binder kinase activator like 1B|Mob1A|MOB4B|Mob4B protein|MOBK1B|Mobkl1b|MOL1B_HUMAN|Mps one binder kinase activator like 1B|Mps one binder kinase activator-like 1B|Protein Mob4B
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MOBKL1B
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MOB1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat