Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-OR10A4 Rabbit Polyclonal Antibody
Anti-OR10A4 Rabbit Polyclonal Antibody
Catalog # 77737-540
Supplier:  AFG Bioscience
New Product
Anti-OR10A4 Rabbit Polyclonal Antibody
Catalog # 77737-540
Supplier:  AFG Bioscience
Supplier Number:  A22351-50UG
New Product

Specifications

  • Antigen Symbol:
    OR10A4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    Q9H209
  • Antigen Synonyms:
    OR10A4P|Olfactory receptor 10A4|HP2|Olfactory receptor-like protein JCG5
  • Storage Buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Olfactory receptor-like protein JCG5
  • Purification:
    Antigen affinity purified
  • Packaging:
    Vial

Specifications

About this item

Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:40000

Type:
Antigen: OR10A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human