Anti-HOXD12 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOXD12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-602
- 77740-322
- Antigen Symbol:
- HOXD12
- Antigen Synonyms:
- Homeobox protein Hox D12|Homeobox protein Hox-4H|Homeobox protein Hox-D12|Hox 4.7|mouse|homolog of|Hox 4H|HOX4H|HOXD12|HXD12_HUMAN
- Homeobox protein Hox D12|Homeobox protein Hox-4H|Homeobox protein Hox-D12|Hox 4.7|mouse|homolog of|Hox 4H|HOX4H|HOXD12|HXD12_HUMAN
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P35452
- P35452
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-D12 protein (30-169AA)
- Recombinant Human Homeobox protein Hox-D12 protein (30-169AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice