Anti-HOXA5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOXA5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-287
- 77857-569
- Antigen Symbol:
- HOXA5
- HOXA5
- Antigen Synonyms:
- Homeo box 1C|Homeo box A5|Homeobox A5|Homeobox protein Hox-1C|Homeobox protein Hox-A5|Homeobox protein HOXA5|HOX 1|HOX 1.3|HOX 1C|HOX1|HOX1.3|HOX1C|HOXA5|HXA5_HUMAN|MGC9376|OTTHUMP00000217259
- Homeo box 1C|Homeo box A5|Homeobox A5|Homeobox protein Hox-1C|Homeobox protein Hox-A5|Homeobox protein HOXA5|HOX 1|HOX 1.3|HOX 1C|HOX1|HOX1.3|HOX1C|HOXA5|HXA5_HUMAN|MGC9376|OTTHUMP00000217259
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20719
- P20719
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-A5 protein (1-270AA)
- Recombinant Human Homeobox protein Hox-A5 protein (1-270AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice