Anti-NKTR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NKTR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-470
- 77744-469
- Antigen Symbol:
- NKTR
- NKTR
- Antigen Synonyms:
- Natural killer triggering receptor|Natural killer tumor recognition sequence|Natural-killer cells cyclophilin-related protein|NK-TR protein|NK-tumor recognition protein|NKTR_HUMAN|p104
- Natural killer triggering receptor|Natural killer tumor recognition sequence|Natural-killer cells cyclophilin-related protein|NK-TR protein|NK-tumor recognition protein|NKTR_HUMAN|p104
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30414
- P30414
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NK-tumor recognition protein (10-175AA)
- NK-tumor recognition protein (10-175AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice