Anti-MRI1 Rabbit Polyclonal Antibody
77743-386
New Product
- Antigen Symbol:MRI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q9BV20
- Antigen Synonyms:EI2BL|M1Pi|Mediator of RhoA-dependent invasion|Methylthioribose 1 phosphate isomerase 1|Methylthioribose 1 phosphate isomerase homolog|Methylthioribose-1-phosphate isomerase|MGC3207|MRDI|mri1|MTNA|MTNA_HUMAN|MTR-1-P isomerase|S methyl 5 thioribose 1 phosphate isomerase 1|S-methyl-5-thioribose-1-phosphate isomerase|Translation initiation factor eIF 2B|Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein|Ypr118w
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Ypr118w
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MRI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat