Anti-MED31 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: MED31
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-632
- 77742-969
- Antigen Symbol:
- MED31
- MED31
- Antigen Synonyms:
- SOH1|CGI-125|Mediator of RNA polymerase II transcription subunit 31|Mediator complex subunit 31|Mediator complex subunit SOH1|hSOH1
- SOH1| CGI-125| Mediator of RNA polymerase II transcription subunit 31| Mediator complex subunit 31| Mediator complex subunit SOH1| hSOH1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3C7
- Q9Y3C7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mediator of RNA polymerase II transcription subunit 31 protein (2-72AA)
- Recombinant Human Mediator of RNA polymerase II transcription subunit 31 protein (2-72AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice