Anti-JAM2 Rabbit Polyclonal Antibody
77741-391
New Product
- Antigen Symbol:JAM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P57087
- Antigen Synonyms:C21orf43| CD 322| CD322| CD322 antigen| JAM 2| JAM B| JAM IT/VE JAM| JAM| vascular endothelial| JAM-2| JAM-B| JAM2_HUMAN| JAMB| Jcam2| Junction adhesion molecule 2| Junction adhesion molecule B| Junctional adhesion molecule 2| Junctional adhesion molecule B| PRO 245| PRO245| Vascular endothelial junction associated molecule| Vascular endothelial junction-associated molecule| VE JAM| VE-JAM| VEJAM
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:VEJAM
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: JAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat