Anti-KPNA1 Rabbit Polyclonal Antibody
77741-813
New Product
- Antigen Symbol:KPNA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:P52294
- Antigen Synonyms:IMA5_HUMAN|Importin alpha 1 subunit|Importin alpha 5|Importin alpha S1|Importin subunit alpha-5|N-terminally processed|IPO A5|IPOA 5|IPOA5|Karyopherin alpha 1|Karyopherin alpha 1 subunit|Karyopherin subunit alpha-1|KPNA 1|KPNA1|mSRP 1|mSRP1|NPI 1|NPI-1|NPI1|Nucleoprotein interactor 1|RAG cohort protein 2|RCH 2|RCH2|Recombination activating gene cohort 2|SRP 1|SRP1 beta|SRP1-beta
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:SRP1-beta
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: KPNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat