Anti-MARK3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MARK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-487
- 77742-788
- Antigen Symbol:
- MARK3
- MARK3
- Antigen Synonyms:
- C-TAK1|Cdc25C associated protein kinase 1|Cdc25C-associated protein kinase 1|cTAK1|ELKL motif kinase 2|EMK-2|Emk2|ETK 1|KIAA4230|KP78|MAP microtubule affinity regulating kinase 3|MAP/microtubule affinity-regulating kinase 3|MARK3_HUMAN|Par 1a|PAR1A|Protein kinase STK10|Ser/Thr protein kinase PAR-1|Serine threonine protein kinase p78|Serine/threonine-protein kinase p78|SerThr protein kinase PAR 1
- C-TAK1|Cdc25C associated protein kinase 1|Cdc25C-associated protein kinase 1|cTAK1|ELKL motif kinase 2|EMK-2|Emk2|ETK 1|KIAA4230|KP78|MAP microtubule affinity regulating kinase 3|MAP/microtubule affinity-regulating kinase 3|MARK3_HUMAN|Par 1a|PAR1A|Protein kinase STK10|Ser/Thr protein kinase PAR-1|Serine threonine protein kinase p78|Serine/threonine-protein kinase p78|SerThr protein kinase PAR 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27448
- P27448
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human MAP/microtubule affinity-regulating kinase 3 protein (368-667AA)
- Recombinant Human MAP/microtubule affinity-regulating kinase 3 protein (368-667AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice