Anti-HTR2A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HTR2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-517
- 77857-750
- Antigen Symbol:
- HTR2A
- Antigen Synonyms:
- 5 HT 2|5 HT 2A|5 HT2 receptor|5 HT2A|5 hydroxytryptamine receptor 2A|5-HT-2|5-HT-2A|5-hydroxytryptamine (serotonin) receptor 2A|G protein-coupled|5-hydroxytryptamine 2A receptor|5-hydroxytryptamine receptor 2A|5HT2A_HUMAN|HTR 2|HTR 2A|HTR2|formerly|HTR2A|serotonin 5-HT-2 receptor|serotonin 5-HT-2A receptor|Serotonin receptor 2A
- 5 HT 2|5 HT 2A|5 HT2 receptor|5 HT2A|5 hydroxytryptamine receptor 2A|5-HT-2|5-HT-2A|5-hydroxytryptamine (serotonin) receptor 2A|G protein-coupled|5-hydroxytryptamine 2A receptor|5-hydroxytryptamine receptor 2A|5HT2A_HUMAN|HTR 2|HTR 2A|HTR2|formerly|HTR2A|serotonin 5-HT-2 receptor|serotonin 5-HT-2A receptor|Serotonin receptor 2A
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P28223
- P28223
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human 5-hydroxytryptamine receptor 2A protein (385-471AA)
- Recombinant Human 5-hydroxytryptamine receptor 2A protein (385-471AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice