Anti-GUCY2C Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GUCY2C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-623
- 77739-283
- Antigen Symbol:
- GUCY2C
- GUCY2C
- Antigen Synonyms:
- GC-C|Guanylyl cyclase C|GUC2C|GUC2C_HUMAN|Heat stable enterotoxin receptor|Heat-stable enterotoxin receptor|hSTAR|Intestinal guanylate cyclase|STA receptor
- GC-C|Guanylyl cyclase C|GUC2C|GUC2C_HUMAN|Heat stable enterotoxin receptor|Heat-stable enterotoxin receptor|hSTAR|Intestinal guanylate cyclase|STA receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25092
- P25092
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Heat-stable enterotoxin receptor protein (636-891AA)
- Recombinant Human Heat-stable enterotoxin receptor protein (636-891AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice