Anti-TAS2R45 Rabbit Polyclonal Antibody
Catalog # 77737-053
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TAS2R45
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:P59539
- Antigen Synonyms:TAS2R45|GPR59|Taste receptor type 2 member 45|T2R45|G-protein coupled receptor 59
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:G-protein coupled receptor 59
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:5000
Type:
Antigen: TAS2R45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human