Anti-GTF2A2 Rabbit Polyclonal Antibody
Catalog # 77739-211
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GTF2A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P52657
- Antigen Synonyms:General transcription factor IIA|General transcription factor IIA subunit 2|2 (12kD subunit)|2|12kDa|gtf2a2|HsT18745|T2AG_HUMAN|TF2A2|TFIIA 12|TFIIA|TFIIA gamma|TFIIA p12 subunit|TFIIA-12|TFIIA-gamma|TFIIA12|TFIIAS|Transcription initiation factor IIA gamma chain|Transcription initiation factor IIA subunit 2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Transcription initiation factor IIA subunit 2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GTF2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat