Anti-GRM7 Rabbit Polyclonal Antibody
Catalog # 77737-677
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GRM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q14831
- Antigen Synonyms:FLJ40498|GLUR7|Glutamate receptor metabotropic 7|GPRC1G|GRM7_HUMAN|Metabotropic glutamate receptor 7|MGLU7|mGluR7|OTTHUMP00000206961|OTTHUMP00000214674|OTTHUMP00000214675
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:FLJ40498
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:40000
Type:
Antigen: GRM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat