Anti-IL13 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IL13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-943
- 77858-009
- Antigen Symbol:
- IL13
- IL13
- Antigen Synonyms:
- Allergic rhinitis|ALRH|BHR 1|BHR1|Bronchial hyperresponsiveness 1 (bronchial asthma)|IL 13|IL-13|IL13_HUMAN|Interleukin 13|Interleukin-13|interleukin13|MGC116786|MGC116788|MGC116789|NC 30|NC30|P 600|P600
- Allergic rhinitis|ALRH|BHR 1|BHR1|Bronchial hyperresponsiveness 1 (bronchial asthma)|IL 13|IL-13|IL13_HUMAN|Interleukin 13|Interleukin-13|interleukin13|MGC116786|MGC116788|MGC116789|NC 30|NC30|P 600|P600
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35225
- P35225
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interleukin-13 protein (30-125AA)
- Recombinant Human Interleukin-13 protein (30-125AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice