Anti-NUP98 Rabbit Recombinant Polyclonal Antibody
77745-236
New Product
- Antigen Symbol:NUP98
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P52948
- Antigen Synonyms:96 kDa nucleoporin|98 kDa nucleoporin|ADAR2|ADIR2|GLFG-repeat containing nucleoporin|Nuclear pore complex protein Nup96|Nuclear pore complex protein Nup98 Nup96|Nucleoporin 98kD|nucleoporin 98kDa|Nucleoporin Nup96|Nucleoporin NUP196|NUP96|Nup98-Nup96|NUP98_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Nuclear pore complex protein Nup98-Nup96 protein (965-1110AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IF: 1:50 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: NUP98
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human