Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-INPP5B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-INPP5B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-INPP5B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77858-125
  • 77741-101
  • Antigen Symbol:
  • INPP5B
  • Antigen Synonyms:
  • 4|5-trisphosphate 5-phosphatase|5PTase|75 kDa inositol polyphosphate 5 phosphatase|75 kDa inositol polyphosphate-5-phosphatase|75kDa|EC 3.1.3.36|I5P2_HUMAN|Inositol polyphosphate 5-phosphatase II|Inositol polyphosphate-5-phosphatase B|Inositol polyphosphate-5-phosphatase|INPP5B|Inpp5b protein|INPP5P|MGC65156|MGC71303|OTTHUMP00000004726|OTTHUMP00000004727|OTTHUMP00000004728|Phosphoinositide 5 phosphatase|Phosphoinositide 5-phosphatase|Type II inositol 1|5 trisphosphate 5 phosphatase|Type II inositol-1
  • 4|5-trisphosphate 5-phosphatase|5PTase|75 kDa inositol polyphosphate 5 phosphatase|75 kDa inositol polyphosphate-5-phosphatase|75kDa|EC 3.1.3.36|I5P2_HUMAN|Inositol polyphosphate 5-phosphatase II|Inositol polyphosphate-5-phosphatase B|Inositol polyphosphate-5-phosphatase|INPP5B|Inpp5b protein|INPP5P|MGC65156|MGC71303|OTTHUMP00000004726|OTTHUMP00000004727|OTTHUMP00000004728|Phosphoinositide 5 phosphatase|Phosphoinositide 5-phosphatase|Type II inositol 1|5 trisphosphate 5 phosphatase|Type II inositol-1
  • Clonality:
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Gene ID:
  • P32019
  • P32019
  • Isotype:
  • IgG
  • ELISA:
  • Yes
  • Immunogen:
  • Recombinant Human Type II inositol 1,4,5-trisphosphate 5-phosphatase protein (78-178AA)
  • Recombinant Human Type II inositol 1,4,5-trisphosphate 5-phosphatase protein (78-178AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications