Anti-FKBP14 Rabbit Polyclonal Antibody
77737-226
New Product
- Antigen Symbol:FKBP14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NWM8
- Antigen Synonyms:22 kDa FK506 binding protein|22 kDa FK506-binding protein|22 kDa FKBP|FK506 binding protein 14 (22 kDa)|FK506 binding protein 14|FK506-binding protein 14|FKB14_HUMAN|FKBP 22|FKBP-14|FKBP-22|FKBP14|FKBP22|FLJ20731|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP14|PPIase|PPIase FKBP14|Rotamase
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Rotamase
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: FKBP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat