Anti-IFNA5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: IFNA5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-892
- 77740-687
- Antigen Symbol:
- IFNA5
- IFNA5
- Antigen Synonyms:
- IeIF G|IFN alphaG|IFN-alpha-5|IFN-AlphaG|IFNA5_HUMAN|INA5|INFA5|Interferon alpha 61|Interferon alpha G|Interferon alpha-5|Interferon alpha-61|Interferon alpha-G|interferon|alpha 5|LeIF G
- IeIF G| IFN alphaG| IFN-alpha-5| IFN-AlphaG| IFNA5_HUMAN| INA5| INFA5| Interferon alpha 61| Interferon alpha G| Interferon alpha-5| Interferon alpha-61| Interferon alpha-G| interferon| alpha 5| LeIF G
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01569
- P01569
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human Interferon alpha-5 protein (52-73AA)
- Peptide sequence from Human Interferon alpha-5 protein (52-73AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice