Anti-NDUFV3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFV3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-253
- 77744-254
- Antigen Symbol:
- NDUFV3
- NDUFV3
- Antigen Synonyms:
- CI-10k|CI-9kD|complex I 10kDa subunit|complex I|mitochondrial respiratory chain|10-kD subunit|Complex I-9kD|mitochondrial NADH oxidoreductase-like protein|NADH dehydrogenase [ubiquinone] flavoprotein 3|mitochondrial|NADH ubiquinone oxidoreductase 9 kD subunit|NADH-ubiquinone oxidoreductase 9 kDa subunit|NADH-ubiquinone oxidoreductase flavoprotein 3|10kD|NDUV3_HUMAN|Renal carcinoma antigen NY-REN-4
- CI-10k|CI-9kD|complex I 10kDa subunit|complex I|mitochondrial respiratory chain|10-kD subunit|Complex I-9kD|mitochondrial NADH oxidoreductase-like protein|NADH dehydrogenase [ubiquinone] flavoprotein 3|mitochondrial|NADH ubiquinone oxidoreductase 9 kD subunit|NADH-ubiquinone oxidoreductase 9 kDa subunit|NADH-ubiquinone oxidoreductase flavoprotein 3|10kD|NDUV3_HUMAN|Renal carcinoma antigen NY-REN-4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P56181
- P56181
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NADH dehydrogenase [ubiquinone] flavoprotein 3, mitochondrial protein (35-108AA)
- NADH dehydrogenase [ubiquinone] flavoprotein 3, mitochondrial protein (35-108AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together