Anti-MPP1 Rabbit Polyclonal Antibody
77743-330
New Product
- Antigen Symbol:MPP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q00013
- Antigen Synonyms:55 kDa erythrocyte membrane protein|AAG 12|AAG12|Aging associated gene 12|DXS552|DXS552E|EM55_HUMAN|EMP 55|EMP55|Erythrocyte membrane protein p55|Membrane protein|Membrane protein palmitoylated 1 55kDa|Membrane protein palmitoylated 1|palmitoylated 1|55kDa|Migration related gene 1|MPP 1|MRG 1|MRG1|OTTHUMP00000026054|OTTHUMP00000196176|p55|Palmitoylated erythrocyte membrane protein|Palmitoylated membrane protein 1|PEMP
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:PEMP
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat