Anti-PCLAF Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PCLAF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-567
- 77858-498
- Antigen Symbol:
- PCLAF
- PCLAF
- Antigen Synonyms:
- HCV NS5A transactivated protein 9|HCV NS5A-transactivated protein 9|Hepatitis C virus NS5A transactivated protein 9|Hepatitis C virus NS5A-transactivated protein 9|KIAA0101|L5|NS5 ATP9|NS5ATP9|OEATC 1|OEATC|OEATC-1|OEATC1|Overexpressed in anaplastic thyroid carcinoma 1|p15(PAF)|p15PAF|PAF|PAF_HUMAN|PCNA associated factor|PCNA-associated factor
- HCV NS5A transactivated protein 9|HCV NS5A-transactivated protein 9|Hepatitis C virus NS5A transactivated protein 9|Hepatitis C virus NS5A-transactivated protein 9|KIAA0101|L5|NS5 ATP9|NS5ATP9|OEATC 1|OEATC|OEATC-1|OEATC1|Overexpressed in anaplastic thyroid carcinoma 1|p15(PAF)|p15PAF|PAF|PAF_HUMAN|PCNA associated factor|PCNA-associated factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15004
- Q15004
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human PCNA-associated factor protein (1-111AA)
- Recombinant Human PCNA-associated factor protein (1-111AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice