Anti-GRIN2C Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRIN2C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-037
- 77856-461
- Antigen Symbol:
- GRIN2C
- GRIN2C
- Antigen Synonyms:
- GluN2C|Glutamate [NMDA] receptor subunit epsilon-3|Glutamate Receptor Ionotropic N Methyl D Aspartate 2C|Glutamate receptor ionotropic, NMDA 2C|Glutamate receptor, ionotropic, N methyl D aspartate 2C|N Methly D Aspartate Receptor Channel Subunit Epsilon 3|N Methyl D Aspartate Receptor Subtype 2C|N methyl D aspartate receptor subunit 2C|N-methyl D-aspartate receptor subtype 2C|NMDAR2C|NMDE3_HUMAN|NR2C|OTTHUMP00000041930
- GluN2C|Glutamate [NMDA] receptor subunit epsilon-3|Glutamate Receptor Ionotropic N Methyl D Aspartate 2C|Glutamate receptor ionotropic, NMDA 2C|Glutamate receptor, ionotropic, N methyl D aspartate 2C|N Methly D Aspartate Receptor Channel Subunit Epsilon 3|N Methyl D Aspartate Receptor Subtype 2C|N methyl D aspartate receptor subunit 2C|N-methyl D-aspartate receptor subtype 2C|NMDAR2C|NMDE3_HUMAN|NR2C|OTTHUMP00000041930
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14957
- Q14957
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Glutamate receptor ionotropic, NMDA 2C protein (908-1007AA)
- Recombinant human Glutamate receptor ionotropic, NMDA 2C protein (908-1007AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice