Anti-NDUFB2 Rabbit Polyclonal Antibody
Catalog # 77744-171
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NDUFB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O95178
- Antigen Synonyms:CI-AGGG|Complex I-AGGG|mitochondrial|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 2|NADH-ubiquinone oxidoreductase AGGG subunit|NDUB2_HUMAN|NDUFB2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:CI-AGGG
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NDUFB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat