Anti-OSGIN1 Rabbit Polyclonal Antibody
Catalog # 77745-714
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OSGIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UJX0
- Antigen Synonyms:BDGI|OKL38|Oxidative stress-induced growth inhibitor 1|Bone marrow stromal cell-derived growth inhibitor|BMSC-derived growth inhibitor|Ovary|kidney and liver protein 38|huOKL38|Pregnancy-induced growth inhibitor OKL38
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Pregnancy-induced growth inhibitor OKL38
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: OSGIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat