Anti-LAMB1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LAMB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-965
- 77858-823
- Antigen Symbol:
- LAMB1
- LAMB1
- Antigen Synonyms:
- CLM|Cutis laxa with marfanoid phenotype|LAM B1|LAMB 1|LAMB1_HUMAN|Laminin B1|Laminin B1 chain|Laminin beta 1 chain|Laminin beta 1 chain precursor|Laminin beta1|Laminin subunit beta 1|Laminin subunit beta-1|Laminin-1 subunit beta|Laminin-10 subunit beta|Laminin-12 subunit beta|Laminin-2 subunit beta|Laminin-6 subunit beta|Laminin-8 subunit beta|LIS5|MGC142015
- CLM|Cutis laxa with marfanoid phenotype|LAM B1|LAMB 1|LAMB1_HUMAN|Laminin B1|Laminin B1 chain|Laminin beta 1 chain|Laminin beta 1 chain precursor|Laminin beta1|Laminin subunit beta 1|Laminin subunit beta-1|Laminin-1 subunit beta|Laminin-10 subunit beta|Laminin-12 subunit beta|Laminin-2 subunit beta|Laminin-6 subunit beta|Laminin-8 subunit beta|LIS5|MGC142015
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07942
- P07942
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Laminin subunit beta-1 protein (523-661AA)
- Recombinant Human Laminin subunit beta-1 protein (523-661AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice