Anti-MCHR2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MCHR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-577
- 77742-889
- Antigen Symbol:
- MCHR2
- MCHR2
- Antigen Synonyms:
- G protein coupled receptor 145|G protein coupled receptor slt|G-protein coupled receptor 145|GPR145|GPRv17|MCH receptor 2|MCH-2R|MCH-R2|MCH2|MCH2R|MCHR-2|MCHR2_HUMAN|Melanin concentrating hormone receptor 2|Melanin-concentrating hormone receptor 2|SLT
- G protein coupled receptor 145|G protein coupled receptor slt|G-protein coupled receptor 145|GPR145|GPRv17|MCH receptor 2|MCH-2R|MCH-R2|MCH2|MCH2R|MCHR-2|MCHR2_HUMAN|Melanin concentrating hormone receptor 2|Melanin-concentrating hormone receptor 2|SLT
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969V1
- Q969V1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Melanin-concentrating hormone receptor 2 protein (1-39AA)
- Recombinant Human Melanin-concentrating hormone receptor 2 protein (1-39AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice