Anti-TAB2 Rabbit Polyclonal Antibody
Catalog # 77742-689
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TAB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NYJ8
- Antigen Synonyms:CHTD2|FLJ21885|KIAA0733|MAP3K7IP2|Mitogen activated protein kinase kinase kinase 7 interacting protein 2|Mitogen-activated protein kinase kinase kinase 7-interacting protein 2|OTTHUMP00000040125|TAB 2|TAB-2|Tab2|TAB2_HUMAN|TAK1 binding protein 2|TAK1-binding protein 2|TGF beta activated kinase 1/MAP3K7 binding protein 2|TGF-beta-activated kinase 1 and MAP3K7-binding protein 2|TGF-beta-activated kinase 1-binding protein 2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MAP3K7IP2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat