Anti-IFT88 Rabbit Polyclonal Antibody
77740-735
New Product
- Antigen Symbol:IFT88
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13099
- Antigen Synonyms:D13S1056E|DAF19|hTg737|IFT88_HUMAN|Intraflagellar transport 88 homolog|Intraflagellar transport protein 88 homolog|MGC26259|Polaris homolog|Probe hTg737 (polycystic kidney disease|autosomal recessive)|Recessive polycystic kidney disease protein Tg737 homolog|RP11-172H24.2|Tetratricopeptide repeat domain 10|Tetratricopeptide repeat protein 10|TG737|TPR repeat protein 10|TTC10
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TTC10
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: IFT88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat