Anti-CNMD Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CNMD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-886
- 77742-056
- Antigen Symbol:
- CNMD
- CNMD
- Antigen Synonyms:
- BRICD3|BRICHOS domain containing 3|CH-SP|CHM I|ChM-I|CHM1|chondromodulin|Chondromodulin I|Chondromodulin-1|Chondromodulin-I|Chondrosurfactant protein|LECT 1|Lect1|LECT1_HUMAN|Leukocyte cell derived chemotaxin 1|Multiple myeloma tumor suppressor 1|MYETS1
- BRICD3|BRICHOS domain containing 3|CH-SP|CHM I|ChM-I|CHM1|chondromodulin|Chondromodulin I|Chondromodulin-1|Chondromodulin-I|Chondrosurfactant protein|LECT 1|Lect1|LECT1_HUMAN|Leukocyte cell derived chemotaxin 1|Multiple myeloma tumor suppressor 1|MYETS1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75829
- O75829
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Leukocyte cell-derived chemotaxin 1 protein (75-334AA)
- Recombinant human Leukocyte cell-derived chemotaxin 1 protein (75-334AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice