Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GRPR Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-GRPR Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-GRPR Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77856-495
  • 77739-083
  • Antigen Symbol:
  • GRPR
  • GRPR
  • Antigen Synonyms:
  • BB2| Bombesin BB2 receptor| bombesin receptor 2| Gastrin Releasing Peptide Receptor| Gastrin-releasing peptide receptor| GRP preferring bombesin receptor| GRP R| GRP-preferring bombesin receptor| GRP-R| Grp/bombesin receptor| Grpr| GRPR_HUMAN
  • BB2|Bombesin BB2 receptor|bombesin receptor 2|Gastrin Releasing Peptide Receptor|Gastrin-releasing peptide receptor|GRP preferring bombesin receptor|GRP R|GRP-preferring bombesin receptor|GRP-R|Grp/bombesin receptor|Grpr|GRPR_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P30550
  • P30550
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Gastrin-releasing peptide receptor protein (326-384AA)
  • Recombinant Human Gastrin-releasing peptide receptor protein (326-384AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications