Anti-GBA Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GBA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-831
- 77738-022
- Antigen Symbol:
- GBA
- GBA
- Antigen Synonyms:
- Acid beta glucosidase|Acid beta-glucosidase|Alglucerase|Beta glucocerebrosidase|BETA GLUCOSIDASE, ACID|Beta-glucocerebrosidase|betaGC|D glucosyl N acylsphingosine glucohydrolase|D-glucosyl-N-acylsphingosine glucohydrolase|EC 3.2.1.45|Gba protein|GBA1|GC|GCase|GCB|GLCM_HUMAN|GLUC|Glucocerebrosidase (alt.)|Glucocerebrosidase|GLUCOCEREBROSIDASE PSEUDOGENE|Glucosidase beta|Glucosidase, beta, acid|Glucosidase, beta, acid (includes glucosylceramidase)|Glucosylceramidase|Imiglucerase|Lysosomal glucocerebrosidase|OTTHUMP00000033992|OTTHUMP00000033993
- Acid beta glucosidase|Acid beta-glucosidase|Alglucerase|Beta glucocerebrosidase|BETA GLUCOSIDASE, ACID|Beta-glucocerebrosidase|betaGC|D glucosyl N acylsphingosine glucohydrolase|D-glucosyl-N-acylsphingosine glucohydrolase|EC 3.2.1.45|Gba protein|GBA1|GC|GCase|GCB|GLCM_HUMAN|GLUC|Glucocerebrosidase (alt.)|Glucocerebrosidase|GLUCOCEREBROSIDASE PSEUDOGENE|Glucosidase beta|Glucosidase, beta, acid|Glucosidase, beta, acid (includes glucosylceramidase)|Glucosylceramidase|Imiglucerase|Lysosomal glucocerebrosidase|OTTHUMP00000033992|OTTHUMP00000033993
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04062
- P04062
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Glucosylceramidase protein (215-343AA)
- Recombinant human Glucosylceramidase protein (215-343AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together