Anti-ADGRL2 Rabbit Polyclonal Antibody
Catalog # 77736-782
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ADGRL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Form:Liquid
- Gene ID:O95490
- Antigen Synonyms:Adgrl2|Adhesion G protein-coupled receptor L2|AGRL2_HUMAN|Calcium-independent alpha-latrotoxin receptor 2|CIRL-2|CL2|KIAA0786|Latrophilin 1|Latrophilin 2|Latrophilin homolog 1|latrophilin homolog 2|Latrophilin-2|LEC1|Lectomedin-1|LPH2|LPHH1|LPHN2_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:LPHN2_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:100 to 1:300, IF:1:200 to 1:1000, ELISA:1:5000
Type:
Antigen: ADGRL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat