Anti-FOXO4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-460
- 77855-573
- Antigen Symbol:
- FOXO4
- FOXO4
- Antigen Synonyms:
- AFX|AFX1|Afxh|ALL1-fused gene from X chromosome|Fork head domain transcription factor AFX1|Forkhead box O4|Forkhead box protein O4|FOXO 4|FOXO4_HUMAN|MGC117660|MGC120490|Mixed lineage leukemia|translocated to|7|MLLT7|Myeloid/lymphoid or mixed lineage leukemia (trithorax homolog|Drosophila)|Myeloid/lymphoid or mixed lineage leukemia|RGD1561201
- AFX|AFX1|Afxh|ALL1-fused gene from X chromosome|Fork head domain transcription factor AFX1|Forkhead box O4|Forkhead box protein O4|FOXO 4|FOXO4_HUMAN|MGC117660|MGC120490|Mixed lineage leukemia|translocated to|7|MLLT7|Myeloid/lymphoid or mixed lineage leukemia (trithorax homolog|Drosophila)|Myeloid/lymphoid or mixed lineage leukemia|RGD1561201
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P98177
- P98177
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human Forkhead box protein O4 protein (45-63AA)
- Peptide sequence from Human Forkhead box protein O4 protein (45-63AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice